Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CKAP5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501986762
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-6762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-6762 Supplier Bioss Supplier No. BS5738R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Colonic and hepatic tumor over-expressed protein (ch-TOG) belongs to the TOG/XMAP215/Dis1 evolutionarily ancient family of microtubule-associated proteins with isoforms found in all major kingdoms of eukaryotes. This protein is required for the assembly and function of the meiotic or mitotic spindles during cell division and is localized to microtubule-organizing centers (MTOCs) during interphase or to the spindle poles and centrosomes during mitosis or cytokinesis.

Specifications

Antigen CKAP5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CKAP5
Gene Accession No. Q14008
Gene Alias 3110043H24Rik; 4930432B04Rik; CHTOG; Ch-TOG; ch-TOGp; CKAP5; Colonic and hepatic tumor overexpressed gene protein; colonic and hepatic tumor over-expressed gene protein; cytoskeleton associated protein 5; Cytoskeleton-associated protein 5; D730027C18Rik; KIAA0097; mKIAA0097; MSPS; T25636; TOG; TOGp
Gene Symbols CKAP5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CKAP5/ch TOG.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9793
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.