Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CKB Mouse anti-Human, Clone: 2B7C3, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | CKB |
---|---|
Clone | 2B7C3 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-7170
![]() ![]() |
Proteintech Group Inc
667641IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized.Specifications
CKB | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B CK, CKB, CKBB, Creatine kinase B chain, Creatine kinase B type, creatine kinase, brain | |
CKB | |
IgG2a | |
Protein A | |
Antibody |
2B7C3 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P12277 | |
1152 | |
CKB Fusion Protein Ag7933 | |
Primary | |
-20°C | |
CKB |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title