Learn More
CKI-gamma1 Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304360A

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine
Casein kinase I (also designated CKI) and casein kinase II (also designated CKII) compose a family of serine/ threonine protein kinases which are present in all eukaryotes examined to date. CKI family members, which include CKI alpha, gamma, epsilon and delta, have been implicated in the control of cytoplasmic and nuclear processes, including DNA replication and repair. CKII is usually expressed as a tetrameric complex consisting of either an alpha2 beta2 or an alpha alpha′ beta2 structure. The alpha catalytic subunit is stimulated by the beta regulatory subunit, which undergoes autophosphorylation. CKII activity is high in the cytosol and nucleus of proliferating and differentiating cells. CKII is known to phosphorylate more than 100 different substrates including nuclear oncoproteins, transcription factors and enzymes involved in DNA metabolism.Specifications
CKI-gamma1 | |
Polyclonal | |
Unconjugated | |
CSNK1G1 | |
Casein kinase I isoform gamma-1, CKI-gamma 1, CSNK1G1, KC1G1, CK1G1, CKI gamma1, CK-I gamma1, CKI- gamma1, CKI gamma-1, CKI gamma1, CK I gamma1, CKI gamma1, CKI gamma 1, CKI gamma 1, CKI-gamma-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
214897, 53944 | |
4° C | |
Liquid |
Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8BTH8, Q9HCP0 | |
Csnk1g1 | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.