Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CKLF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595787
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595787 Supplier Invitrogen™ Supplier No. PA595787
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDNVQPKIKH RPFCFSVKGH VKMLRLVFAL VTAVCCLADG ALIYRKLLFN PSGPYQKKPV HEKKEVL; Positive Samples: Mouse thymus, Mouse testis, Rat thymus, Rat testis; Cellular Location: Membrane, Multi-pass membrane protein.

CKLF is a cytokine. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 16. CKLF is a potent chemoattractant for neutrophils, monocytes and lymphocytes. It also can stimulate the proliferation of skeletal muscle cells. This protein may play important roles in inflammation and in the regeneration of skeletal muscle. The product of this gene is a cytokine. Cytokines are small proteins that have an essential role in the immune and inflammatory responses. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 16. The protein encoded by this gene is a potent chemoattractant for neutrophils, monocytes and lymphocytes. It also can stimulate the proliferation of skeletal muscle cells. This protein may play important roles in inflammation and in the regeneration of skeletal muscle. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKLF
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CKLF
Gene Accession No. Q9DAS1, Q9JK79
Gene Alias 1700001C14Rik; 1810018M11Rik; C32; Chemokine-like factor; chemokine-like factor 1; chemokine-like factor 2; chemokine-like factor 3; chemokine-like factor 4; chemokine-like factor 6; CKLF; CKLF1; Cklf2; CKLF3; CKLF4; CKLF5; Cklf6; HSPC224; transmembrane proteolipid; UCK-1; UNQ410/PRO772
Gene Symbols CKLF
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-67 of human CKLF (NP_0574101).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245978, 75458
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.