Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CKLF Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984687
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4687 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4687 Supplier Bioss Supplier No. BS2394R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CKLF is a cytokine. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 16. CKLF is a potent chemoattractant for neutrophils, monocytes and lymphocytes. It also can stimulate the proliferation of skeletal muscle cells. This protein may play important roles in inflammation and in the regeneration of skeletal muscle. The product of this gene is a cytokine. Cytokines are small proteins that have an essential role in the immune and inflammatory responses. This gene is one of several chemokine-like factor genes located in a cluster on chromosome 16. The protein encoded by this gene is a potent chemoattractant for neutrophils, monocytes and lymphocytes. It also can stimulate the proliferation of skeletal muscle cells. This protein may play important roles in inflammation and in the regeneration of skeletal muscle. Alternatively spliced transcript variants encoding different isoforms have been identified.

Specifications

Antigen CKLF
Applications Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CKLF
Gene Accession No. Q9UBR5
Gene Alias 1700001C14Rik; 1810018M11Rik; C32; Chemokine-like factor; chemokine-like factor 1; chemokine-like factor 2; chemokine-like factor 3; chemokine-like factor 4; chemokine-like factor 6; Cklf; Cklf1; CKLF2; CKLF3; CKLF4; CKLF5; Cklf6; HSPC224; transmembrane proteolipid; UCK-1; UNQ410/PRO772
Gene Symbols CKLF
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CKLF.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51192
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.