Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CKLFSF2 isoform 1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980188
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0188 Supplier Bioss Supplier No. BS0265R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene belongs to the chemokine-like factor gene superfamily, a novel family that links the chemokine and the transmembrane 4 superfamilies of signaling molecules. The protein encoded by this gene may play an important role in testicular development.

Specifications

Antigen CKLFSF2 isoform 1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene CMTM2
Gene Accession No. Q8TAZ6
Gene Alias 1700001K04Rik; 1700041N15Rik; 1700063K20Rik; AI449770; ARR19; BOS_17102; C32; chemokine-like factor super family 1; chemokine-like factor super family 2A; chemokine-like factor superfamily 2; chemokine-like factor superfamily 2-1b; chemokine-like factor superfamily 2-like; Chemokine-like factor superfamily member 2; Chemokine-like factor superfamily member 2A; Cklf; CKLF like MARVEL transmembrane domain containing 2; Cklf1; CKLF3; CKLF4; CKLF5; CKLF-like MARVEL transmembrane domain containing 2; CKLF-like MARVEL transmembrane domain containing 2A; CKLF-like MARVEL transmembrane domain-containing protein 2; CKLF-like MARVEL transmembrane domain-containing protein 2A; Cklfsf1; CKLFSF2; Cklfsf2-1b; Cklfsf2a; CMTM2; Cmtm2a; LOC100342033; UCK-1
Gene Symbols CMTM2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mo CKLFSF2 isoform 1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 146225
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.