Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CKMT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5111344
Description
Immunogen sequence: ASIFSKLLTG RNASLLFATM GTSVLTTGYL LNRQKVCAEV REQPRLFPPS ADYPDL.
Mitochondrial creatine kinase (MtCK) is responsible for the transfer of high energy phosphate from mitochondria to the cytosolic carrier, creatine. It belongs to the creatine kinase isoenzyme family. It exists as two isoenzymes, sarcomeric MtCK and ubiquitous MtCK, encoded by separate genes. Mitochondrial creatine kinase occurs in two different oligomeric forms: dimers and octamers, in contrast to the exclusively dimeric cytosolic creatine kinase isoenzymes. Sarcomeric mitochondrial creatine kinase has 80% homology with the coding exons of ubiquitous mitochondrial creatine kinase. This gene contains sequences homologous to several motifs that are shared among some nuclear genes encoding mitochondrial proteins and thus may be essential for the coordinated activation of these genes during mitochondrial biogenesis. Three transcript variants encoding the same protein have been found for this gene.
Specifications
| CKMT2 | |
| Polyclonal | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P17540 | |
| Ckmt2 | |
| Recombinant Protein corresonding to Human CKMT2. Recombinant protein control fragment (Product #RP-101289). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Ckmt2 | |
| 2300008A19Rik; basic-type mitochondrial creatine kinase; CKMT2; Creatine kinase S-type, mitochondrial; creatine kinase, mitochondrial 2; creatine kinase, mitochondrial 2 (sarcomeric); creatine kinase, mitochondrial 2, sarcomeric; EC 2.7.3.2; mib-CK; sarcomeric mitochondrial creatine kinase; ScCKmit; SMTCK; S-MtCK | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 1160 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction