Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CKS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582090
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582090 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582090 Supplier Invitrogen™ Supplier No. PA582090
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HKQIYYSDKY FDEHYEYRHV MLPRELSKQV PKTHLMSEEE WRRLGVQQSL GWVHYMIHEP EPHILLFRRP LPKDQQ Highest antigen sequence indentity to the following orthologs: Mouse - 99%, Rat - 99%.

CKS2 protein binds to the catalytic subunit of the cyclin dependent kinases and is essential for their biological function. The CKS2 mRNA is found to be expressed in different patterns through the cell cycle in HeLa cells, which reflects specialized role for the encoded protein. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKS2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cks2
Gene Accession No. P33552
Gene Alias 1110038L14Rik; CDC28 protein kinase 2; CDC28 protein kinase regulatory subunit 2; CKS1(S. cerevisiae Cdc28/Cdc2 kinase subunit) homolog-2; CKS2; CKS-2; CKSHS2; Cyclin-dependent kinases regulatory subunit 2; RGD1562047; RP11-8C21.2
Gene Symbols Cks2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CKS2. Recombinant protein control fragment (Product #RP-101912).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1164
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.