Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CL-K1/COLEC11 Antibody (148), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290474PCP
Description
CL-K1/COLEC11 Monoclonal antibody specifically detects CL-K1/COLEC11 in Mouse samples. It is validated for ELISASpecifications
CL-K1/COLEC11 | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
148 | |
ELISA | |
CLK1, CL-K1, CL-K1-I, CL-K1-II, CL-K1-IIa, CL-K1-IIb, collectin kidney I, Collectin kidney protein 1, collectin sub-family member 11, collectin-11, DKFZp686N1868, MGC3279 | |
Rabbit immunized with purified, recombinant Mouse CL-K1/COLEC11 (Uniprot#: Q3SXB8-1, Met1-Leu272) | |
0.1 mL | |
Cell Biology | |
78989 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction