Learn More
Invitrogen™ CLAC-P Polyclonal Antibody

Description
PA1-4307 detects CLAC-P (NC2-2 region) in human samples. PA1-4307 has been successfully used in Western blot procedures. By Western blot this antibody detects bands of 50, 70, and 100 kDa representing CLAC-P in formic acid extracts of Alzheimer's disease brain. PA1-4307 immunizing peptide corresponds to amino acid residues 155-169 of the NC2-2 region of human and mouse CLAC-P.
Specifications
Specifications
| Antigen | CLAC-P |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | COL25A1 |
| Gene Accession No. | Q9BXS0 |
| Gene Alias | 2700062B08Rik; Alzheimer disease amyloid-associated protein; AMY; CFEOM5; CLAC; CLACP; CLAC-P; Col25a1; collagen alpha-1(XXV) chain; collagen type XXV alpha 1; collagen, type XXV, alpha 1; collagen-like Alzheimer amyloid plaque component; collagenous Alzheimer amyloid plaque component; procollagen, type XXV, alpha 1 |
| Gene Symbols | COL25A1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.