Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLAC-P Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14308
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14308 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14308 Supplier Invitrogen™ Supplier No. PA14308
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4308 detects CLAC-P (NC3 region) in human and mouse samples. PA1-4308 has been successfully used in Western blotting and immunohistochemistry procedures. By Western blot, this antibody detects a ~70 kDa protein representing CLAC-P (NC3 region) in stable HEK293T cell lines expressing CLAC-P (NC3 region). PA1-4308 does not cross react with COLXIII or COLXXIII proteins. PA1-4308 immunizing peptide corresponds to amino acid residues 430-443 of the NC3 region of human and mouse CLAC-P. Maintain at 4ºC for 3 months or at -20ºC for longer periods. Avoid freeze/thaw cycles.

Alzheimer's disease (AD) is characterized by deposition of beta-amyloid as senile plaques and cerebral amyloid angiopathy. The principal component of AD amyloid is the amyloid beta peptide (A beta). Although production and deposition of A beta appear closely related to the pathogenesis of AD, current evidence suggest that A beta alone is not sufficient to cause neuronal death and symptoms of dementia. A component of senile plaque (SP), CLAC (collagen-like Alzheimer amyloid plaque component ) and its precursor CLAC-P are deposited with extracellular beta-amyloid and have been recently implicated in cell toxicity of beta-amyloid in AD brains. CLAC-P is a unique, membrane-bound collagen-like structure containing three Gly-X-Y repeat motifs and may define a novel class of neuronal collagens.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLAC-P
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene COL25A1
Gene Accession No. Q99MQ5, Q9BXS0
Gene Alias 2700062B08Rik; Alzheimer disease amyloid-associated protein; AMY; CFEOM5; CLAC; CLACP; CLAC-P; Col25a1; collagen alpha-1(XXV) chain; collagen type XXV alpha 1; collagen, type XXV, alpha 1; collagen-like Alzheimer amyloid plaque component; collagenous Alzheimer amyloid plaque component; procollagen, type XXV, alpha 1
Gene Symbols COL25A1
Host Species Rabbit
Immunogen Synthetic peptide: D(430) Y N G N L H E A L Q R I T(443) of the NC3 region of human and murine CLAC-P.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 77018, 84570
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.