Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLACP/COL25A1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981544
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1544 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1544 Supplier Bioss Supplier No. BS11643R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Alzheimer’s disease (AD) is characterized by deposition of beta-amyloid as senile plaques and cerebral amyloid angiopathy. The principal component of AD amyloid is the amyloid beta peptide (A beta). Although production and deposition of A beta appear closely related to the pathogenesis of AD, current evidence suggest that A beta alone is not sufficient to cause neuronal death and symptoms of dementia. A component of senile plaque (SP), CLAC (collagen-like Alzheimer amyloid plaque component ) and its precursor CLAC-P are deposited with extracellular beta-amyloid and have been recently implicated in cell toxicity of beta-amyloid in AD brains. CLAC-P is a unique, membrane-bound collagen-like structure containing three Gly-X-Y repeat motifs and may define a novel class of neuronal collagens.

Specifications

Antigen CLACP/COL25A1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene COL25A1
Gene Accession No. Q9BXS0
Gene Alias 2700062B08Rik; Alzheimer disease amyloid-associated protein; AMY; CFEOM5; CLAC; CLACP; CLAC-P; COL25A1; Collagen alpha-1(XXV) chain; collagen type XXV alpha 1; collagen, type XXV, alpha 1; Collagen-like Alzheimer amyloid plaque component; collagenous Alzheimer amyloid plaque component; procollagen, type XXV, alpha 1
Gene Symbols COL25A1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human CLACP/COL25A1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84570
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.