Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clarithromycin, 960-1040 ^mg/mg

Catalog No. p-7057445
Click to view available options
Quantity:
100 mg
1 g
5 g

CAS: 81103-11-9 | C38H69NO13 | 747.964 g/mol

A macrolide antibiotic that inhibits bacterial growth by targeting the 50S ribosomal subunit

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A macrolide antibiotic that inhibits bacterial growth by targeting the 50S ribosomal subunit
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81103-11-9
Molecular Formula C38H69NO13
Molecular Weight (g/mol) 747.964
MDL Number MFCD00865140
InChI Key AGOYDEPGAOXOCK-KCBOHYOISA-N
PubChem CID 84029
ChEBI CHEBI:3732
IUPAC Name (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-12,13-dihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dion
SMILES CCC1C(C(C(C(=O)C(CC(C(C(C(C(C(=O)O1)C)OC2CC(C(C(O2)C)O)(C)OC)C)OC3C(C(CC(O3)C)N(C)C)O)(C)OC)C)C)O)(C)O

Specifications

Type Clarithromycin
Physical Form Powder
Melting Point 217°C to 220°C
Color White
Quantity 100 mg
Flash Point >110°C (230°F)
Boiling Point 86°C
Merck Index 14,2339
Solubility Information Soluble in DMSO
Formula Weight 747.95
Chemical Name or Material Clarithromycin, 960 to 1040μg/mg
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
RTECSNumber KF4997000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.