Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLASP1 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP119127R
Description
CLASP1 Polyclonal antibody specifically detects CLASP1 in Human samples. It is validated for Western BlotSpecifications
CLASP1 | |
Polyclonal | |
Western Blot | |
CLIP-associating protein 1, cytoplasmic linker associated protein 1, Cytoplasmic linker-associated protein 1, DKFZp686D1968, DKFZp686H2039, hOrbit1, KIAA0622FLJ33821, MAST1FLJ41222, MGC131895, multiple asters 1, Multiple asters homolog 1, Protein Orbit homolog 1 | |
The immunogen recognized by this antibody maps to a region between residue 1488 and 1538 of human cytoplasmic linker associated protein 1 using the numbering given in entry NP_056097.1 (GeneID 23332). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
23332 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction