Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLASP1 Antibody (KT66), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP323219
Description
CLASP1 Monoclonal antibody specifically detects CLASP1 in Human, Mouse, Rat, Chicken, Xenopus samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
CLASP1 | |
Monoclonal | |
Unconjugated | |
PBS | |
Rat | |
Protein G purified | |
RUO | |
23332 | |
Store at 4°C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Western Blot, Immunofluorescence | |
KT66 | |
Western Blot 1:500, Immunocytochemistry/Immunofluorescence 1:100 | |
CLIP-associating protein 1, cytoplasmic linker associated protein 1, Cytoplasmic linker-associated protein 1, DKFZp686D1968, DKFZp686H2039, hOrbit1, KIAA0622FLJ33821, MAST1FLJ41222, MGC131895, multiple asters 1, Multiple asters homolog 1, Protein Orbit homolog 1 | |
Clone KT66 was generated from a GST fusion protein containing the C-terminus of mouse CLASP1. This sequence is highly conserved in human, rat, chicken, and fish CLASP1, and has low homology to CLASP2. [UniProt# Q80TV8] | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Chicken, Xenopus species | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction