Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLASP1 Antibody (SR1237), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | CLASP1 |
---|---|
Clone | SR1237 |
Dilution | Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Description
CLASP1 Monoclonal antibody specifically detects CLASP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
CLASP1 | |
Western Blot 1:1000-1:5000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
23332 | |
IgG | |
Affinity purified |
SR1237 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
CLIP-associating protein 1, cytoplasmic linker associated protein 1, Cytoplasmic linker-associated protein 1, DKFZp686D1968, DKFZp686H2039, hOrbit1, KIAA0622FLJ33821, MAST1FLJ41222, MGC131895, multiple asters 1, Multiple asters homolog 1, Protein Orbit homolog 1 | |
A synthesized peptide derived from human CLASP1 (Uniprot #: Q7Z460) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title