Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLASP1 Monoclonal Antibody (KT66)
Rat Monoclonal Antibody
Supplier: Invitrogen™ MA547708
Description
The antibody detects a 161 kDa* protein corresponding to the molecular mass of CLASP1 on SDS-PAGE immunoblots of human HeLa, mouse brain, and rat PC12 cells. The antibody also detects CLASP1 by immunocytochemistry in HeLa cells.
CLASPs, such as CLASP1, are nonmotor microtubule-associated proteins that interact with CLIPs.
Specifications
CLASP1 | |
Monoclonal | |
~1.0 mg/mL | |
PBS with 0.05% sodium azide | |
Q7Z460, Q80TV8 | |
CLASP1 | |
Clone KT66 was generated from a GST fusion protein containing the C-terminus of mouse CLASP1. This sequence is highly conserved in human, rat, chicken, and fish CLASP1, and has low homology to CLASP2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2a |
Western Blot, Immunocytochemistry | |
KT66 | |
Unconjugated | |
CLASP1 | |
1700030C23Rik; 5730583A19Rik; B130045P17Rik; CLASP1; CLASP1alpha; CLIP associating protein 1; CLIP-associating protein 1; cytoplasmic linker associated protein 1; cytoplasmic linker-associated protein 1; hOrbit1; Kiaa0622; MAST1; multiple asters 1; multiple asters homolog 1; Protein Orbit homolog 1 | |
Rat | |
Protein G | |
RUO | |
23332, 304740, 76707 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction