Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLASP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121064
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121064 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121064 Supplier Invitrogen™ Supplier No. PA5121064
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HepG2, MCF7, Mouse brain, Rat brain. The target is usually found in the following locations: Chromosome, Cytoplasm, Golgi apparatus, centromere, centrosome, cytoskeleton, kinetochore, microtubule organizing center, spindle, trans-Golgi network. Immunogen sequence: IMKTLEAHKD SHKEVVRAAE EAASTLASSI HPEQCIKVLC PIIQTADYPI NLAAIKMQTK VVERIAKESL LQLLVDIIPG LLQGYDNTES SVRKASVFCL VAIYSVIGED LKPHLAQLTG SKMKLLNLYI KRAQTTNSNS SSSSDVSTHS.

CLASPs, such as CLASP1, are nonmotor microtubule-associated proteins that interact with CLIPs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLASP1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CLASP1
Gene Accession No. Q7Z460, Q80TV8
Gene Alias 1700030C23Rik; 5730583A19Rik; B130045P17Rik; CLASP1; CLASP1alpha; CLIP associating protein 1; CLIP-associating protein 1; cytoplasmic linker associated protein 1; cytoplasmic linker-associated protein 1; hOrbit1; Kiaa0622; MAST1; multiple asters 1; multiple asters homolog 1; Protein Orbit homolog 1
Gene Symbols CLASP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1389-1538 of human CLASP1 (NP_056097.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23332, 304740, 76707
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.