Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLASP1 Recombinant Rabbit Monoclonal Antibody (JE43-19)
GREENER_CHOICE

Catalog No. PIMA534834
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA534834 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA534834 Supplier Invitrogen™ Supplier No. MA534834
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Control: Daudi, HepG2, SH-SY-5Y, SiHa, rat kidney tissue, human appendix tissue, human small intestine tissue, mouse colon tissue.

CLASPs, such as CLASP1, are nonmotor microtubule-associated proteins that interact with CLIPs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLASP1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone JE43-19
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene CLASP1
Gene Accession No. Q7Z460
Gene Alias 1700030C23Rik; 5730583A19Rik; B130045P17Rik; CLASP1; CLASP1alpha; CLIP associating protein 1; CLIP-associating protein 1; cytoplasmic linker associated protein 1; cytoplasmic linker-associated protein 1; hOrbit1; Kiaa0622; MAST1; multiple asters 1; multiple asters homolog 1; Protein Orbit homolog 1
Gene Symbols CLASP1
Host Species Rabbit
Immunogen Synthetic peptide within Human CLASP1 aa 1-50.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23332
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.