Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLASP2 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP121394R
Description
CLASP2 Polyclonal antibody specifically detects CLASP2 in Human samples. It is validated for Western BlotSpecifications
CLASP2 | |
Polyclonal | |
Western Blot | |
CLIP-associating protein 2, cytoplasmic linker associated protein 2, Cytoplasmic linker-associated protein 2, hOrbit2, KIAA0627, multiple asters (Mast)-like homolog 2, Protein Orbit homolog 2 | |
The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human cytoplasmic linker associated protein 2 using the numbering given in entry BAG11221.1 (GeneID 23122). | |
0.1 mL | |
Cell Biology, Cell Cycle and Replication | |
23122 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction