Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLASP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103750 Supplier Invitrogen™ Supplier No. PA5103750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CLASP2.

CLASP2 is microtubule plus-end tracking protein that promotes the stabilization of dynamic microtubules. Required for the polarization of the cytoplasmic microtubule arrays in migrating cells towards the leading edge of the cell. May act at the cell cortex to enhance the frequency of rescue of depolymerizing microtubules by attaching their plus-ends to cortical platforms composed of ERC1 and PHLDB2. This cortical microtubule stabilizing activity is regulated at least in part by phosphatidylinositol 3-kinase signaling. Also performs a similar stabilizing function at the kinetochore which is essential for the bipolar alignment of chromosomes on the mitotic spindle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLASP2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Clasp2
Gene Accession No. O75122
Gene Alias 1500004F14Rik; 8030404L10Rik; C77448; CLASP2; CLASP2alpha; CLASP2beta; CLASP2gamma; CLIP associating protein 2; CLIP-associating protein 2; CLIP-associating protein CLASP2; cytoplasmic linker associated protein 2; cytoplasmic linker-associated protein 2; hOrbit2; Kiaa0627; mKIAA0627; multiple asters (Mast)-like homolog 2; protein Orbit homolog 2
Gene Symbols Clasp2
Host Species Rabbit
Immunogen A synthesized peptide derived from human CLASP2(Accession O75122), corresponding to amino acid residues S313-G363.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23122
Target Species Human, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.