Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CLASP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109547
Description
Immunogen sequence: NCTSKSVPVR RRSFEFLDLL LQEWQTHSLE RHAAVLVETI KKGIHDADAE ARVEARKTYM GLRNHFPGEA ETLYNSLEPS YQKSLQTYLK SSGSVASLPQ SDRSSSSSQE SLNRPFSSKW STANPSTVAG RVSAGSSKAS SLPGSLQRSR SDIDVNAAAG AKAHHAAGQS VRSGRLGAGA L.
CLASP2 is microtubule plus-end tracking protein that promotes the stabilization of dynamic microtubules. Required for the polarization of the cytoplasmic microtubule arrays in migrating cells towards the leading edge of the cell. May act at the cell cortex to enhance the frequency of rescue of depolymerizing microtubules by attaching their plus-ends to cortical platforms composed of ERC1 and PHLDB2. This cortical microtubule stabilizing activity is regulated at least in part by phosphatidylinositol 3-kinase signaling. Also performs a similar stabilizing function at the kinetochore which is essential for the bipolar alignment of chromosomes on the mitotic spindle.
Specifications
| CLASP2 | |
| Polyclonal | |
| Unconjugated | |
| Clasp2 | |
| 1500004F14Rik; 8030404L10Rik; C77448; CLASP2; CLASP2alpha; CLASP2beta; CLASP2gamma; CLIP associating protein 2; CLIP-associating protein 2; CLIP-associating protein CLASP2; cytoplasmic linker associated protein 2; cytoplasmic linker-associated protein 2; hOrbit2; Kiaa0627; mKIAA0627; multiple asters (Mast)-like homolog 2; protein Orbit homolog 2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114514, 23122, 76499 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.04 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75122, Q8BRT1, Q99JD4 | |
| Clasp2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 220-400 of human CLASP2 (NP_001193973.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction