Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claspin Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100248R
Description
Claspin Polyclonal antibody specifically detects Claspin in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
Claspin | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
claspin, claspin homolog, claspin homolog (Xenopus laevis), hClaspin, MGC131612, MGC131613, MGC131615 | |
The immunogen recognized by this antibody maps to a region between residues 900 and 950 of human Claspin using the numbering given in entry AAG24515.1 (GeneID 63967). | |
0.1 mL | |
Cancer, Cell Cycle and Replication, DNA Repair | |
63967 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction