Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claspin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 4 publications
Supplier: Novus Biologicals NB100248
Description
Claspin Polyclonal specifically detects Claspin in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
Claspin | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
claspin, claspin homolog, claspin homolog (Xenopus laevis), hClaspin, MGC131612, MGC131613, MGC131615 | |
Rabbit | |
180 kDa | |
100 μL | |
Cancer, Cell Cycle and Replication, DNA Repair | |
63967 | |
Human, Mouse | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
1.0 mg/mL | |
Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 2-10 ug/mg of lysate | |
AAG24515.1 | |
CLSPN | |
The immunogen recognized by this antibody maps to a region between residues 900 and 950 of human Claspin using the numbering given in entry AAG24515.1 (GeneID 63967). | |
Affinity Purified | |
RUO | |
Primary | |
Human. Based on 100% sequence identity, this antibody is predicted to react with Gorilla and Chimpanzee. | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction