Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Claspin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582909
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582909 Supplier Invitrogen™ Supplier No. PA582909
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SFVILEPETN RELEALKQRF WKHANPAAKP RAGQTVNVNV IVKDMGTDGK EELKADVVPV TLAPKKLDGA SHTKPGEKLQ VLKAKLQEAM KLRR Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 87%.

Xenopus claspin is an essential upstream regulator of checkpoint kinase 1 and triggers a checkpoint arrest of the cell cycle in the presence of DNA templates in Xenopus egg extracts. The human gene appears to be the homolog Xenopus claspin and its function has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claspin
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLSPN
Gene Accession No. Q9HAW4
Gene Alias B130025E01; C85083; claspin; claspin homolog; CLSPN; E130314M08Rik; hClaspin; MGC131612; MGC131613; MGC131615
Gene Symbols CLSPN
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CLSPN. Recombinant protein control fragment (Product #RP-105764).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63967
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.