Learn More
Claspin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300265AT
Description
The recommended shelf life for this product is 1 year from date of receipt.
Xenopus claspin is an essential upstream regulator of checkpoint kinase 1 and triggers a checkpoint arrest of the cell cycle in the presence of DNA templates in Xenopus egg extracts. The human gene appears to be the homolog Xenopus claspin and its function has not been determined.Specifications
| Claspin | |
| Polyclonal | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| Q9HAW4 | |
| CLSPN | |
| Between 450 and 500 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| CLSPN | |
| CLASPIN, MGC131612, MGC131613, MGC131615 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 63967 | |
| 4° C | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.