Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLASRP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566494
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566494 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566494 Supplier Invitrogen™ Supplier No. PA566494
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PAVGEKLKKTE PAAGKETGAA KPKLTPQEKL KLRMQKALNR QFKADKKAAQ EKMIQQEHER QEREDELRAM Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.

CLASRP is probably functions as an alternative splicing regulator. CLASRP may regulate the mRNA splicing of genes such as CLK1. This may act by regulating members of the CLK kinase family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLASRP
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CLASRP
Gene Accession No. Q8N2M8
Gene Alias Clasp; Clasrp; CLK4 associating serine/arginine rich protein; Clk4 associating SR-related protein; CLK4-associating serine/arginine rich protein; Clk4-associating SR-related protein; Serine/arginine-rich splicing factor 16; Sfrs16; Splicing factor, arginine/serine-rich 16; splicing factor, arginine/serine-rich 16 (suppressor-of-white-apricot homolog, Drosophila); Srsf16; suppressor of white apricot homolog 2; suppressor of white-apricot homolog 2; SWAP2; UNQ2428/PRO4988
Gene Symbols CLASRP
Host Species Rabbit
Immunogen Recombinant Human CLASRP. Recombinant protein control fragment (Product #RP-107165).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11129
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.