Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BINDER™ Class 3.3 Independent Temperature Safety Device
Shop All BINDER Inc. Products

Click to view available options
For Use With (Equipment):
BINDER Chambers KBF 240
Description
- Class 3.3
- With visual alarm
- Safety devices also available in Class 2 and 3.1
Specifications
Specifications
Description | Class 3.3 independent temperature safety device - with visual alarm (DIN 12880) |
For Use With (Equipment) | BINDER Chambers KBF 240 |
Material | Plastic |
Type | Temperature Safety Device |
For Use With (Application) | Temperature Regulation |
Includes | Visual alarm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction