Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hand2Mind Classroom Makerspace Cart Grade

Click to view available options
Grade:
3 to 5
K to 2
Description
- Foster creativity and innovation with engineering concepts using our new Makerspace Carts.
- They're an affordable, fully loaded, turnkey makerspace solution for classrooms, STEM labs, and libraries.
Specifications
Specifications
Grade Level | Elementary |
Grade | K to 2 |
Class Size | 1 Class |
Product Type | Classroom Makerspace Cart |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction