Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hand2Mind Classroom Makerspace Cart

Click to view available options
Class Size:
1 Class
Description
- These colorful carts are designed specifically for a teacher for in a classroom.
- The 9 rainbow colored drawers are perfect for storing and organizing material for a makerspace or just organizing your classroom.
Specifications
Specifications
| Grade Level | Elementary, Intermediate - Middle School, High School |
| Grade | K to 12 |
| Class Size | 1 Class |
| Product Type | Classroom Makerspace Cart |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction