Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clathrin Heavy Chain 1/CHC17 Antibody (CHC/1432), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308333F
Description
Clathrin Heavy Chain 1/CHC17 Monoclonal antibody specifically detects Clathrin Heavy Chain 1/CHC17 in Human, Mouse, Rat, Porcine, Avian, Bovine, Canine, Monkey, Xenopus samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
Clathrin Heavy Chain 1/CHC17 | |
Monoclonal | |
FITC | |
CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034 | |
Recombinant full-length human Clathrin Heavy Chain 1/CHC17 protein | |
100 μg | |
Membrane Trafficking and Chaperones, Membrane Vesicle Markers | |
1213 | |
Store at 4°C in the dark. | |
IgG1 κ |
Immunofluorescence | |
CHC/1432 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Pig, Avian | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction