Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin Heavy Chain 1/CHC17 Antibody (CLTC/1431) - Azide and BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$610.00

Specifications

Antigen Clathrin Heavy Chain 1/CHC17
Clone CLTC/1431
Concentration 1 mg/mL
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, CyTOF-ready
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB254300100
SDP
View Documents
Novus Biologicals
NBP254300100UG
100 μg
Each for $610.00
Only null left
Add to Cart
 
NB254300200
SDP
View Documents
Novus Biologicals
NBP254300200UG
200 μg N/A N/A N/A
Description

Description

Clathrin Heavy Chain 1/CHC17 Monoclonal specifically detects Clathrin Heavy Chain 1/CHC17 in Human samples. It is validated for ELISA.
Specifications

Specifications

Clathrin Heavy Chain 1/CHC17
1 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, CyTOF
Unconjugated
Mouse
Membrane Trafficking and Chaperones, Membrane Vesicle Markers
PBS with No Preservative
1213
Purified recombinant N-terminal fragment of human Clathrin Heavy Chain (exact sequence is proprietary)
Primary
Store at -20 to -80C. Avoid freeze-thaw cycles.
CLTC/1431
Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, CyTOF-ready
Monoclonal
Purified
RUO
Human
CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034
CLTC
IgG1 κ
Protein A or G purified
192 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.