Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clathrin Heavy Chain 1/CHC17 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299436PE
Description
Clathrin Heavy Chain 1/CHC17 Polyclonal antibody specifically detects Clathrin Heavy Chain 1/CHC17 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Clathrin Heavy Chain 1/CHC17 | |
Polyclonal | |
Western Blot, Immunohistochemistry-Paraffin | |
CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Clathrin Heavy Chain 1/CHC17 | |
0.1 mL | |
Membrane Trafficking and Chaperones, Membrane Vesicle Markers | |
1213 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction