Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clathrin Heavy Chain 1/CHC17 Antibody (X22), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB300613MFV500
Description
Clathrin Heavy Chain 1/CHC17 Monoclonal antibody specifically detects Clathrin Heavy Chain 1/CHC17 in Human, Mouse, Rat, Bovine, Hamster, Primate samples. It is validated for Flow CytometrySpecifications
Clathrin Heavy Chain 1/CHC17 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Mouse, Rat, Bovine, Hamster, Primate | |
Purified |
Flow Cytometry | |
X22 | |
Flow Cytometry | |
CHC, CHC17, clathrin heavy chain 1, Clathrin heavy chain on chromosome 17, clathrin, heavy chain, clathrin, heavy chain (Hc), clathrin, heavy polypeptide (Hc), clathrin, heavy polypeptide-like 2, CLH17, CLH-17, CLTCL2, Hc, KIAA0034 | |
Purified human brain clathrin heavy chain. | |
0.1 mL | |
Membrane Trafficking and Chaperones, Membrane Vesicle Markers | |
1213 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction