Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin Heavy Chain 2/CHC22/CLTCL1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. p-7248556 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB169231 25 μg
NB169230 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB169231 Supplier R&D Systems Supplier No. NBP31707725UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Clathrin Heavy Chain 2/CHC22/CLTCL1 Polyclonal specifically detects Clathrin Heavy Chain 2/CHC22/CLTCL1 in Human samples. It is validated for Immunohistochemistry∣Immunohistochemistry (Paraffin).

Specifications

Antigen Clathrin Heavy Chain 2/CHC22/CLTCL1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias CHC22, clathrin heavy chain 2, clathrin heavy chain on chromosome 22, clathrin, heavy chain-like 1, Clathrin, heavy polypeptide D, CLTCL, heavy polypeptide-like 1
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: LDALESLRKQEEHVTEPAPLVFDFDGHE
Purification Method Affinity purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8218
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.