Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clathrin Heavy Chain Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588534
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588534 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588534 Supplier Invitrogen™ Supplier No. PA588534
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YLRSVQNHNN KSVNESLNNL FITEEDYQAL RTSIDAYDNF DNISLAQRLE KHELIEFRRI AAYLFKGNNR WKQSVELCKK DSLYKDAMQY ASESKDTELA EELLQWFLQE EKRECFGACL FTCYDLLRPD VVLETAWRHN IMDFAMPYFI QVMKEYLTKV DKLDASESLR KEEEQATETQ PIVYGQPQLM LTAGPSVAVP PQAPFGYGYT APPYGQPQPG FGYSM; Positive Samples: HeLa, HepG2, NIH/3T3, Mouse brain, Mouse lung, Rat brain; Cellular Location: Cytoplasmic side, Cytoplasmic vesicle membrane, Melanosome, Membrane, Peripheral membrane protein, coated pit.

Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clathrin Heavy Chain
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.04 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene CLTCL1
Gene Accession No. P11442, P53675, Q00610, Q68FD5
Gene Alias 3110065L21Rik; CHC; CHC17; CHC22; clathrin heavy chain; clathrin heavy chain 1; Clathrin heavy chain 2; clathrin heavy chain like 1; clathrin heavy chain on chromosome 17; Clathrin heavy chain on chromosome 22; clathrin, heavy chain (Hc); clathrin, heavy chain-like 1; clathrin, heavy polypeptide (Hc); Clathrin, heavy polypeptide D; clathrin, heavy polypeptide-like 1; clathrin, heavy polypeptide-like 2; CLH17; CLH-17; CLH22; CLH-22; Cltc; CLTCL; CLTCL1; CLTCL2; CLTD; Hc; KIAA0034; R74732
Gene Symbols CLTC, CLTCL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1451-1675 of human Clathrin heavy chain (NP_0048501).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1213, 54241, 67300, 8218
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.