Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clathrin interactor 1 Antibody (OTI8C1), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271491AF700
Description
Clathrin interactor 1 Monoclonal specifically detects Clathrin interactor 1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Clathrin interactor 1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CLINT1 | |
Recombinant protein fragment corresponding to amino acids 1-290 of human Clathrin interactor 1 (NP_055481) produced in E.coli. | |
0.1 mL | |
9685.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI8C1 | |
Western Blot | |
clathrin interacting protein localized in the trans-Golgi region, clathrin interactor 1, Clathrin-interacting protein localized in the trans-Golgi region, CLINT, ENTHClint, enthoprotin, EPN4Enthoprotin, EPNRepsin 4, Epsin-4, EpsinR, Epsin-related protein, FLJ46753, KIAA0171epsinR | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction