Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clathrin light chain + heavy chain Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169194
Description
Clathrin light chain + heavy chain Polyclonal specifically detects Clathrin light chain + heavy chain in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Clathrin light chain + heavy chain | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
CLTA | |
Synthetic peptides corresponding to CLTA (clathrin, light chain (Lca)) The peptide sequence was selected from the C terminal of CLTA. Peptide sequence KELEEWYARQDEQLQKTKANNRAAEEAFVNDIDESSPGTEWERVARLCDF. | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Zebrafish: 75%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
clathrin light chain A, clathrin, light chain A, LCA, light polypeptide (Lca) | |
Rabbit | |
24 kDa | |
100 μL | |
Membrane Trafficking and Chaperones, Membrane Vesicle Markers | |
1211 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction