Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Clathrin Light Chain Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA519006
Description
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
Specifications
| Clathrin Light Chain | |
| Polyclonal | |
| Unconjugated | |
| CLTB | |
| 2310046E19Rik; AV026556; CHC17; clathrin light chain A; clathrin light chain A S homeolog; clathrin light chain A S homeolog; clathrin, light chain A S homeolog; clathrin light chain B; clathrin, light chain (Lca); clathrin, light chain (Lcb); clathrin, light chain A; clathrin, light chain A2; clathrin, light chain B; clathrin, light polypeptide (Lca); clathrin, light polypeptide (Lcb); clathrin, light polypeptide A b; clathrin, light polypeptide B (light chain B); clathryn light chain (LCA2); CLH17; clta; clta.S; CLTA2; clta-a; cltab; clta-b; CLTB; CLTC; CLTLB; CTLA; CTLB; KIAA0034; lca; LCA1; LCA2; LCA3; Lcb; unnamed protein product; wu:fb61d08; wu:fj48d06; XELAEV_18001213mg; zgc:73358 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 1211, 1212 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P09496, P09497 | |
| CLTA, CLTB | |
| Synthetic peptide sequence (CDFNPKSSKQAKD) corresponding to the internal amino acids of CLTA (aa 218-230). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction