Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clathrin Light Chain Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100037
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100037 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100037 Supplier Invitrogen™ Supplier No. PA5100037
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CLTA.

Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clathrin Light Chain
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CLTB
Gene Accession No. O08585, P08081, P08082, P09496, P09497, Q6IRU5
Gene Alias 2310046E19Rik; AV026556; CHC17; clathrin light chain A; clathrin light chain A S homeolog; clathrin light chain A S homeolog; clathrin, light chain A S homeolog; clathrin light chain B; clathrin, light chain (Lca); clathrin, light chain (Lcb); clathrin, light chain A; clathrin, light chain A2; clathrin, light chain B; clathrin, light polypeptide (Lca); clathrin, light polypeptide (Lcb); clathrin, light polypeptide A b; clathrin, light polypeptide B (light chain B); clathryn light chain (LCA2); CLH17; clta; clta.S; CLTA2; clta-a; cltab; clta-b; CLTB; CLTC; CLTLB; CTLA; CTLB; KIAA0034; lca; LCA1; LCA2; LCA3; Lcb; unnamed protein product; wu:fb61d08; wu:fj48d06; XELAEV_18001213mg; zgc:73358
Gene Symbols CLTA, CLTB
Host Species Rabbit
Immunogen A synthesized peptide derived from human CLTA(Accession P09496), corresponding to amino acid residues G58-E108.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116561, 1211, 1212, 12757, 74325, 83800
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.