Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-1 Antibody (421203), DyLight 650, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB4618W
Description
Claudin-1 Monoclonal antibody specifically detects Claudin-1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Claudin-1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
421203 | |
Flow Cytometry, CyTOF-ready | |
claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1 | |
CHO Chinese hamster ovary cell line transfected with human Claudin-1, Met1-Val211, Accession # O95832 | |
0.1 mL | |
Extracellular Matrix | |
9076 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction