Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-1 Antibody (CL3698), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Claudin-1 |
---|---|
Clone | CL3698 |
Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
Claudin-1 Monoclonal specifically detects Claudin-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Claudin-1 | |
Western Blot 1 μg/mL, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Monoclonal | |
Purified | |
Extracellular Matrix | |
9076 | |
This antibody was developed against a recombinant protein corresponding to amino acids: KTTSYPTPRPYPKPAPSSGKDYV | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL3698 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
claudin 1, claudin-1, CLD1, ILVASCSenescence-associated epithelial membrane protein, SEMP1senescence-associated epithelial membrane protein 1 | |
CLDN1 | |
IgG1 | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title