Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Claudin 1 Monoclonal Antibody (SP128)
GREENER_CHOICE

Catalog No. PIMA516351
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA516351 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA516351 Supplier Invitrogen™ Supplier No. MA516351
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Heat-mediated antigen retrieval is recommended prior to staining, using a 1mM EDTA buffer, pH 8.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is liver, hepatocellular carcinoma or ovarian carcinoma.

Claudin-1 is a member of the transmembrane protein family claudins located in cell-cell tight junctions and it acts as a co-receptor for HCV entry into hepatic cells. Claudins are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Claudin-1 is expressed in most tissues such as bladder, fallopian tube, liver, pancreas, prostate, and skin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone SP128
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.6
Gene CLDN1
Gene Accession No. O95832
Gene Alias AI596271; claudin 1; claudin-1; CLD1; CLDN1; hypothetical protein; ILVASC; SEMP1; Senescence-associated epithelial membrane protein; senescence-associated epithelial membrane protein 1; tight junction protein; UNQ481/PRO944
Gene Symbols CLDN1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminus of human claudin-1 protein.
Purification Method Protein A/G
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9076
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.