Learn More
Epredia™ Lab Vision™ Claudin 1 Rabbit Polyclonal Antibody with BSA and azide
Specifically detect Claudin 1 in human, monkey, mouse, dog and rat samples with Epredia™ Claudin 1, Rabbit Polyclonal Antibody.
Supplier: Epredia™ RB9209P
Description
Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Monkey, Mouse, Dog and Rat. Others not tested.
Epitope: C-terminal
Immunogen: A synthetic peptide derived from the C-terminus of human Claudin 1
Molecular Weight: 22 kDa
Positive Control: T47D cells, Skin or Breast CA
Cellular Localization: Membrane on skin and cytoplasmic and nuclear on breast carcinoma
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Specifications
| Claudin 1 | |
| Polyclonal | |
| 200µg/mL | |
| Rabbit | |
| 1 mL | |
| Cell Adhesion | |
| Canine, Human, Mouse, Non-human Primate, Rat | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Polyclonal | |
| Unconjugated | |
| A synthetic peptide derived from the C-terminus of human Claudin-1 | |
| IVD | |
| Primary | |
| Concentrated |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.