Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Claudin 1 Recombinant Rabbit Monoclonal Antibody (9C3P1)
GREENER_CHOICE

Catalog No. FEMA551271
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA551271 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA551271 Supplier Invitrogen™ Supplier No. MA551271
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: EVQKMRMAVI GGAIFLLAGL AILVATAWYG NRIVQEFYDP MTPVNARYEF GQALFTGWAA ASLCLLGGAL LCCSCPRKTT SYPTPRPYPK PAPSSGKDYV.

Claudin-1 is a member of the transmembrane protein family claudins located in cell-cell tight junctions and it acts as a co-receptor for HCV entry into hepatic cells. Claudins are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Claudin-1 is expressed in most tissues such as bladder, fallopian tube, liver, pancreas, prostate, and skin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9C3P1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene CLDN1
Gene Accession No. O88551, O95832, P56745
Gene Alias AI596271; claudin 1; claudin-1; CLD1; CLDN1; hypothetical protein; ILVASC; SEMP1; Senescence-associated epithelial membrane protein; senescence-associated epithelial membrane protein 1; tight junction protein; UNQ481/PRO944
Gene Symbols CLDN1
Host Species Rabbit
Immunogen Synthetic peptide
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12737, 65129, 9076
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.