Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Claudin 1 Recombinant Rabbit Monoclonal Antibody (BLR094G)

Catalog No. PIMA544380
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA544380 10 μL
PIMA544379 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA544380 Supplier Invitrogen™ Supplier No. MA544380
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.

Claudin-1 is a member of the transmembrane protein family claudins located in cell-cell tight junctions and it acts as a co-receptor for HCV entry into hepatic cells. Claudins are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Claudin-1 is expressed in most tissues such as bladder, fallopian tube, liver, pancreas, prostate, and skin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone BLR094G
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation BBS with 0.1% BSA and 0.09% sodium azide; pH 8.2
Gene CLDN1
Gene Accession No. O95832
Gene Alias AI596271; claudin 1; claudin-1; CLD1; CLDN1; hypothetical protein; ILVASC; SEMP1; Senescence-associated epithelial membrane protein; senescence-associated epithelial membrane protein 1; tight junction protein; UNQ481/PRO944
Gene Symbols CLDN1
Host Species Rabbit
Immunogen A peptide representing a region between residue 161 and the C-terminus (residue 211) of human claudin-1.
Purification Method Purified
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9076
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.