Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin 10 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299148PE
Description
Claudin 10 Polyclonal antibody specifically detects Claudin 10 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
Claudin 10 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
claudin 10, claudin-10, CPETRL3, OSP-L, OSP-like protein | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Claudin 10. | |
0.1 mL | |
Cell Biology, Cytoskeleton Markers, Signal Transduction | |
9071 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction