Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-10b Antibody (394513), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4279J
Description
Claudin-10b Monoclonal antibody specifically detects Claudin-10b in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Claudin-10b | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9071 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, CyTOF | |
394513 | |
Flow Cytometry, CyTOF-ready | |
CLDN10;CPETRL3;HELIX;OSPL;OSP-L | |
NS0 mouse myeloma cell line transfected with human Claudin-10 b Isoform, Met1-Val228, Accession # P78369 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction