Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-11 Antibody (398205R), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4280RW
Description
Claudin-11 Monoclonal antibody specifically detects Claudin-11 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Claudin-11 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
RUO | |
5010 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, CyTOF | |
398205R | |
Flow Cytometry, CyTOF-ready | |
claudin 11, claudin-11, Oligodendrocyte-specific protein, OSPOTMoligodendrocyte transmembrane protein | |
NS0 mouse myeloma cell line transfected with human Claudin-11, Met1-Val207, Accession # O75508 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction