Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin-11 Antibody (398205R), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4280RMFV500
Description
Claudin-11 Monoclonal antibody specifically detects Claudin-11 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| Claudin-11 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 398205R | |
| Flow Cytometry, CyTOF-ready | |
| claudin 11, claudin-11, Oligodendrocyte-specific protein, OSPOTMoligodendrocyte transmembrane protein | |
| NS0 mouse myeloma cell line transfected with human Claudin-11, Met1-Val207, Accession # O75508 | |
| RUO | |
| 5010 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction